Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dodecatungstophosphate hydrate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Phosphotungstic acid sodium salt hydrate

A11575-36 A11575-22 A11575-14 A11575-36 A11575-22 A11575-14
AAAA11575-36EA 793.99 USD InStock
AAAA11575-36 AAAA11575-14 CAAAAA11575-36 CAAAAA11575-14 CAAAAA11575-22 AAAA11575-22
Sodium dodecatungstophosphate hydrate
Phosphotungstic acid sodium salt hydrate


Formula: Na₃O₄₀PW₁₂·H₂O
MW: 2946.12 g/mol (anhy)
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD00151658
CAS Number: 51312-42-6
EINECS: 257-132-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty