Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dibromobutane 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11669-22 A11669-36 A11669-09 A11669-22 A11669-09 A11669-36
AAA11669-36EA 98.7 USD InStock
AAA11669-36 CAAAA11669-22 AAAA11669-09 CAAAA11669-36 CAAAAA11669-09 AAA11669-22
1,4-Dibromobutane 98+%
1,4-Dibromobutane


Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 194…196 °C
Melting Pt: –20 °C
Density: 1.820
Flash Pt: 131 °C(267 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000261
CAS Number: 110-52-1
EINECS: 203-775-5
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty