Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lauric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dodecylic acidDodecanoic acid

A11672-36 A11672-0C A11672-36 A11672-0C A11672-0E A11672-0E
AAAA11672-0EEA 79.99 USD InStock
AAAA11672-0E CAAAAA11672-36 CAAAAA11672-0C AAAA11672-36 AAAA11672-0C CAAAAA11672-0E
Lauric acid 98%
Lauric acid


Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 180 °C/21 mm
Melting Pt: 43…45 °C
Density: 0.883
Flash Pt: 165 °C(329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty