Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfate 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: Diammonium sulfateSulphuric acid diammonium saltDiammonium sulphateSulfuric acid diammonium salt

A11682-0B A11682-0B A11682-0I A11682-0I
AAAA11682-0BEA 51.02 USD InStock
AAAA11682-0B CAAAAA11682-0B AAAA11682-0I CAAAAA11682-0I
Ammonium sulfate 98+%
Ammonium sulphate


Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: ca 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty