Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) oxalate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Oxalic acid tin (II) saltTin (II) ethanedioate

A11689-22 A11689-22 A11689-0E A11689-36 A11689-36 A11689-0E
AAAA11689-22EA 30.99 USD InStock
AAAA11689-22 CAAAAA11689-36 CAAAAA11689-22 AAAA11689-0E CAAAAA11689-0E AAAA11689-36
Tin(II) oxalate 98%
Tin (II) oxalate


Formula: C₂O₄Sn
MW: 206.73 g/mol
Melting Pt: 280 °C (dec.)
Density: 3.56
Storage Temperature: Ambient
MDL Number: MFCD00040678
CAS Number: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: "6.1",III
Merck Index: 13,08865
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty