Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylmorpholine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Ethylmorpholine

A11905-AU A11905-AP A11905-AU A11905-AP
AAAA11905-APEA 50.69 USD InStock
AAAA11905-AP CAAAAA11905-AP AAA11905-AU CAAAA11905-AU
4-Ethylmorpholine 98%
4-Ethylmorpholine


Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 138…139 °C
Melting Pt: –63 °C
Density: 0.908
Flash Pt: 27 °C(80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty