Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Stilbene 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Isostilbene(Z)-1,2-Diphenylethenecis-1,2-Diphenylethylenecis-1,2-Diphenylethene

A11924-14 A11924-03 A11924-14 A11924-06 A11924-03 A11924-06
AAAA11924-06EA 78.29 USD InStock
AAAA11924-06 CAAAAA11924-14 CAAAAA11924-03 CAAAAA11924-06 AAAA11924-14 AAAA11924-03
cis-Stilbene 97%
cis-Stilbene


Formula: (C₆H₅)CHCH(C₆H₅)
MW: 180.25 g/mol
Boiling Pt: 307 °C
Melting Pt: 1…2 °C
Density: 1.04
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004788
CAS Number: 645-49-8
EINECS: 211-445-7
Merck Index: 12,08972
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty