Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethoxybenzaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

A11987-22 A11987-36 A11987-36 A11987-22 A11987-14 A11987-14
AAAA11987-22EA 96.69 USD InStock
CAAAAA11987-22 AAAA11987-22 CAAAAA11987-36 CAAAAA11987-14 AAAA11987-36 AAAA11987-14
4-Ethoxybenzaldehyde 97+%
4-Ethoxybenzaldehyde


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 253…255 °C
Melting Pt: 13…14 °C
Density: 1,08
Flash Pt: <110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003388
CAS Number: 10031-82-0
EINECS: 233-093-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty