Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Chlorobenzene-1-carbothioamide

A12021-14 A12021-22 A12021-22 A12021-18 A12021-18 A12021-14 A12021-09 A12021-09
AAAA12021-18EA 168.99 USD InStock
CAAAAA12021-18 CAAAAA12021-09 AAAA12021-18 AAA12021-22 AAA12021-14 CAAAA12021-14 AAAA12021-09 CAAAA12021-22
4-Chlorothiobenzamide 97%
4-Chlorothiobenzamide


Formula: C₇H₆ClNS
MW: 171.65 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00040956
CAS Number: 2521-24-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty