Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Acetoxy-3-methoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Vanillin acetate4-Formyl-2-methoxyphenyl acetate

A12103-14 A12103-22 A12103-22 A12103-30 A12103-14 A12103-30
AAA12103-22EA 145.99 USD InStock
CAAAA12103-14 CAAAAA12103-30 AAA12103-22 CAAAA12103-22 AAAA12103-30 AAA12103-14
4-Acetoxy-3-methoxybenzaldehyde 98%
4-Acetoxy-3-methoxybenzaldehyde


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003362
CAS Number: 881-68-5
EINECS: 212-920-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty