Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphonoacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12117-09 A12117-09 A12117-14 A12117-14 A12117-22 A12117-22
AAA12117-14EA 161.99 USD InStock
CAAAAA12117-09 AAA12117-14 CAAAA12117-14 AAA12117-22 AAAA12117-09 CAAAA12117-22
Phosphonoacetic acid 98+%
Phosphonoacetic acid


Formula: C₂H₅O₅P
Melting Pt: 139…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00004311
CAS Number: 4408-78-0
EINECS: 224-558-1
UN: 2923
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty