Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium dihydrogen phosphate 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: MonopotassiumphosphatePotassium dihydrogen orthophosphatePotassium phosphate monobasicMonobasic Potassium PhosphatePotassium orthophosphate monobasicPotassium dihydrogenorthophosphate

A12142-0E A12142-0E A12142-36 A12142-36
AAAA12142-0EEA 145.99 USD InStock
AAAA12142-0E CAAAAA12142-36 CAAAAA12142-0E AAAA12142-36
Potassium dihydrogen phosphate 98+%
Potassium dihydrogen phosphate


Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.338
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty