Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

N-Methyl-2-pyrrolidone (NMP) 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

A12260-36 A12260-36 A12260-0E A12260-0C A12260-0E A12260-0C
AAAA12260-36EA 60.28 USD InStock
AAAA12260-36 CAAAAA12260-0E AAAA12260-0C AAAA12260-0E CAAAAA12260-36 CAAAAA12260-0C
N-Methyl-2-pyrrolidone (NMP) 99+%
N-Methyl-2-pyrrolidone (NMP)


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 204 °C
Melting Pt: –24 °C
Density: 1.033
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty