Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateCitric acid tridosium salt dihydrate

A12274-0E A12274-0C A12274-0E A12274-0C A12274-36 A12274-36
AAAA12274-0EEA 102.75 USD InStock
AAAA12274-0E CAAAAA12274-0C CAAAAA12274-36 CAAAAA12274-0E AAAA12274-0C AAAA12274-36
tri-Sodium citrate dihydrate 99%
tri-Sodium citrate dihydrate


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C (–H₂O of crystall.)
Density: 1.76
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty