Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl-2,2,2-trichloroacetimidate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Trichloroacetimidic Acid Benzyl Ester

A12387-14 A12387-06 A12387-14 A12387-06 A12387-18 A12387-30 A12387-30 A12387-18
AAA12387-30EA 844.99 USD InStock
CAAAA12387-18 CAAAAA12387-14 AAA12387-30 AAAA12387-14 CAAAA12387-30 CAAAAA12387-06 AAA12387-18 AAAA12387-06
Benzyl-2,2,2-trichloroacetimidate 98%
Benzyl-2,2,2-trichloroacetimidate


Formula: C₉H₈Cl₃NO
MW: 252.52 g/mol
Boiling Pt: 110…114 °C/0.5 mm
Density: 1.356
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000805
CAS Number: 81927-55-1
EINECS: 617-271-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty