Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl (2,2-diethoxyethyl)phosphonate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diethyl 2,2-diethoxy-ethanephosphonate

A12461-18 A12461-30 A12461-09
AAAA12461-30EA 1689.99 USD InStock
AAAA12461-30 AAAA12461-09 AAAA12461-18
Diethyl (2,2-diethoxyethyl)phosphonate 96%
Diethyl (2,2-diethoxyethyl)phosphonate


Formula: C₁₀H₂₃O₅P
MW: 254.26 g/mol
Boiling Pt: 101…103 °C/8 mm
Density: 1.052
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009236
CAS Number: 7598-61-0
EINECS: 231-504-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty