Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: p-Nitrobenzoyl chloride

A12543-0B A12543-09 A12543-0B A12543-09 A12543-30 A12543-30
AAA12543-0BEA 139.98 USD InStock
AAA12543-0B AAA12543-30 CAAAAA12543-09 CAAAA12543-30 AAAA12543-09 CAAAA12543-0B
4-Nitrobenzoyl chloride 98%
4-Nitrobenzoyl chloride


Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 155 °C/15 mm
Melting Pt: 71…74 °C
Density: 1.53
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: "8",III
Merck Index: 12,06687
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty