Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium hydroxide 40% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

A12626-AP A12626-AE A12626-0F A12626-AE A12626-0F A12626-AP
AAAA12626-APEA 273.85 USD InStock
AAAA12626-AP CAAAAA12626-AP CAAAAA12626-AE AAAA12626-0F AAAA12626-AE CAAAAA12626-0F
Tetrabutylammonium hydroxide 40% (w/w) in methanol
Tetrabutylammonium hydroxide


Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Density: 0.910
Flash Pt: 18 °C(64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty