Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl hydrogen malonate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Mono-Ethyl malonate3-Ethoxy-3-oxopropanoic acid

A12627-18 A12627-09 A12627-30
AAAA12627-30EA 993.99 USD InStock
AAAA12627-30 AAAA12627-18 AAAA12627-09
Ethyl hydrogen malonate 96%
Ethyl hydrogen malonate


Formula: C₅H₈O₄
MW: 132.115 g/mol
Boiling Pt: 106 °C/3 mm
Density: 1.119
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00020490
CAS Number: 1071-46-1
EINECS: 213-992-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty