Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-chloro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A12635-14 A12635-22 A12635-22 A12635-14 A12635-06 A12635-06
AAAA12635-22EA 290.99 USD InStock
CAAAA12635-06 CAAAAA12635-22 AAAA12635-22 CAAAA12635-14 AAA12635-06 AAA12635-14
1-Bromo-4-chloro-2-nitrobenzene 98%
1-Bromo-4-chloro-2-nitrobenzene


Formula: C₆H₃BrClNO₂
MW: 236.45 g/mol
Melting Pt: 68…72 °C
Density: 2.048 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00024320
CAS Number: 41513-04-6
EINECS: 255-421-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty