Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethylcaproic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Ethylhexanoic acid

A12644-AP A12644-0D A12644-AE A12644-0F A12644-0D A12644-0F A12644-AP A12644-AE
AAAA12644-0FEA 87.1 USD InStock
AAAA12644-0F CAAAAA12644-0F AAAA12644-0D AAAA12644-AE CAAAAA12644-0D CAAAAA12644-AE AAAA12644-AP CAAAAA12644-AP
2-Ethylcaproic acid 99%
2-Ethylcaproic acid


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 226…229 °C
Melting Pt: –59 °C
Density: 0.91
Flash Pt: 114 °C(237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002675
CAS Number: 149-57-5
EINECS: 205-743-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty