Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A12648-22 A12648-14 A12648-06 A12648-22 A12648-06 A12648-14
AAAA12648-22EA 486.99 USD InStock
CAAAA12648-14 CAAAAA12648-22 AAAA12648-22 CAAAA12648-06 AAA12648-14 AAA12648-06
4-Chloro-3-nitrobenzonitrile 98%
4-Chloro-3-nitrobenzonitrile


Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00016987
CAS Number: 939-80-0
EINECS: 213-364-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty