Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: PABAVitamin Bxpara-Aminobenzoic acidVitamin H1

A12673-0B A12673-18 A12673-30 A12673-18 A12673-30 A12673-0B
AAAA12673-30EA 85.25 USD InStock
AAAA12673-30 CAAAAA12673-0B AAAA12673-0B CAAAAA12673-18 AAAA12673-18 CAAAAA12673-30
4-Aminobenzoic acid 99%
4-Aminobenzoic acid


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: ca 187 °C (dec.)
Density: 1.374
Storage Temperature: Ambient
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty