Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobutan-1-ol 98%

Supplier: Thermo Scientific Chemicals
Danger

A12680-14 A12680-06 A12680-06 A12680-03 A12680-14 A12680-03
AAAA12680-06EA 126.8 USD InStock
CAAAAA12680-06 AAAA12680-06 CAAAAA12680-14 CAAAAA12680-03 AAAA12680-03 AAAA12680-14
4-Aminobutan-1-ol 98%
4-Aminobutan-1-ol


Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206 °C
Melting Pt: 16…18 °C
Density: 0.967
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008230
CAS Number: 13325-10-5
EINECS: 236-364-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty