Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hippuric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Benzoylaminoacetic acid Benzoyl amidoacetic acid N-Benzoylglycine Benzoyl glycocoll

A12690-36 A12690-0B A12690-22 A12690-36 A12690-0B A12690-22
AAAA12690-22EA 50.64 USD InStock
CAAAAA12690-36 AAAA12690-22 AAA12690-0B AAAA12690-36 CAAAA12690-0B CAAAAA12690-22
Hippuric acid 98%
Hippuric acid


Formula: C₆H₅C(O)NHCH₂COOH
MW: 179.18 g/mol
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 188…191 °C
Density: 1.371
Storage Temperature: Ambient
MDL Number: MFCD00002692
CAS Number: 495-69-2
EINECS: 207-806-3
Merck Index: 13,04735
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty