Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylthiophene 98%

Supplier: Thermo Scientific Chemicals
Danger

A12736-14 A12736-36 A12736-36 A12736-22 A12736-22 A12736-14
AAA12736-14EA 43.49 USD InStock
AAA12736-14 AAAA12736-36 CAAAAA12736-36 CAAAA12736-22 CAAAA12736-14 AAA12736-22
2-Methylthiophene 98%
2-Methylthiophene


Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 112…114 °C
Melting Pt: –63 °C
Density: 1.017
Flash Pt: 7 °C(44 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty