Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium fluoride hydrate ≥87% (dried basis)

Supplier: Thermo Scientific Chemicals
Warning

A12830-36 A12830-0E A12830-36 A12830-0E
AAAA12830-36EA 39.79 USD InStock
CAAAAA12830-36 AAAA12830-36 CAAAAA12830-0E AAAA12830-0E
Magnesium fluoride hydrate ≥87% (dried basis)
Magnesium fluoride hydrate


Formula: F₂Mg·H₂O
MW: 62.30 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty