Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diisopropyl methylphosphonate 95%

Supplier: Thermo Scientific Chemicals
Warning

A12904-14 A12904-06 A12904-14 A12904-22 A12904-22 A12904-06
AAAA12904-14EA 218.99 USD InStock
AAAA12904-14 CAAAAA12904-06 AAAA12904-06 AAAA12904-22 CAAAAA12904-14 CAAAAA12904-22
Diisopropyl methylphosphonate 95%
Diisopropyl methylphosphonate


Formula: C₇H₁₇O₃P
Boiling Pt: 50…51 °C/1 mm
Density: 0.976
Storage Temperature: Ambient
MDL Number: MFCD00015022
CAS Number: 1445-75-6
EINECS: 215-896-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty