Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

A12907-06 A12907-14 A12907-06
AAAA12907-14EA 410.99 USD InStock
AAAA12907-14 CAAAA12907-06 AAA12907-06
4-(Trifluoromethyl)benzenesulfonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride


Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 118…120 °C/18 mm
Melting Pt: 28…34 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042422
CAS Number: 2991-42-6
EINECS: 000-000-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty