Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminoisobutyric acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-2-methylpropanoic acidα-Aminoisobutyric acid2-Amino-2-methylpropionic acid2-Methylalanine2-MetilalaninaH-Aib-OH

A13021-22 A13021-30 A13021-30 A13021-14 A13021-22 A13021-14
AAA13021-30EA 220.99 USD InStock
AAA13021-30 CAAAAA13021-22 CAAAA13021-30 AAAA13021-22 AAAA13021-14 CAAAAA13021-14
2-Aminoisobutyric acid 99%
2-Aminoisobutyric acid


Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008049
CAS Number: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty