Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminopyrazine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Aminopyrazine Pyrazin-2-amine

A13052-14 A13052-18 A13052-06 A13052-14 A13052-06 A13052-18
AAAA13052-14EA 188.99 USD InStock
AAAA13052-14 CAAAAA13052-06 CAAAA13052-18 AAA13052-18 CAAAAA13052-14 AAAA13052-06
2-Aminopyrazine 99%
2-Aminopyrazine


Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty