Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13059-18 A13059-22 A13059-14 A13059-14 A13059-06 A13059-06 A13059-18 A13059-22
AAAA13059-06EA 44.79 USD InStock
CAAAAA13059-22 CAAAAA13059-14 AAAA13059-06 CAAAAA13059-06 CAAAA13059-18 AAAA13059-14 AAAA13059-22 AAA13059-18
2-Iodoaniline 98+%
2-Iodoaniline


Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 53…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty