Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Naphthalene 99+%

Supplier: Thermo Scientific Chemicals
Warning

A13188-0I A13188-30 A13188-0B
AAAA13188-0IEA 181.27 USD InStock
AAAA13188-0I AAAA13188-0B AAAA13188-30
Naphthalene 99+%
Naphthalene


Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218 °C
Melting Pt: 80…82 °C
Density: 0.963
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: "4.1",III
Merck Index: 12,06457
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/18/789118.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty