Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thiosalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Thiobenzoic acid2-Mercaptobenzoic acid

A13401-0B A13401-30 A13401-18 A13401-30 A13401-0B A13401-18
AAA13401-18EA 87.49 USD InStock
CAAAA13401-30 AAA13401-18 CAAAAA13401-0B AAAA13401-0B AAA13401-30 CAAAA13401-18
Thiosalicylic acid 98%
Thiosalicylic acid


Formula: C₇H₆O₂S
MW: 154.19 g/mol
Melting Pt: 165…168 °C
Density: 1.49
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
ADR: 9,
Merck Index: 12,09498
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty