Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrachloro-p-benzoquinone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetrachloro-1,4-benzoquinone Tetrachloroquinone 4-Chloranil p-Chloranil 2,3,5,6-Tetrachloro-1,4-benzoquinone

A13495-36 A13495-22 A13495-14
AAAA13495-36EA 453.99 USD InStock
AAAA13495-36 AAA13495-14 AAA13495-22
Tetrachloro-p-benzoquinone 97%
Tetrachloro-p-benzoquinone


Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: ca 290 °C (subl.)
Density: 1.970
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
UN: 2811
ADR: "6.1",III
Merck Index: 13,02088
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty