Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triiodomethane 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Iodoform

A13664-30 A13664-0B A13664-18 A13664-0B A13664-18 A13664-30
AAAA13664-18EA 42.39 USD InStock
CAAAAA13664-18 AAAA13664-18 CAAAAA13664-30 AAAA13664-30 CAAAAA13664-0B AAAA13664-0B
Triiodomethane 99%
Triiodomethane


Formula: CHI₃
MW: 393.73 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 118…123 °C
Density: 4,008
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Merck Index: 13,05055
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty