Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A13688-30 A13688-0B A13688-36 A13688-0B A13688-30 A13688-36
AAAA13688-36EA 102.99 USD InStock
CAAAA13688-30 AAAA13688-36 AAA13688-30 CAAAAA13688-36 CAAAA13688-0B AAA13688-0B
1,3-Dichlorobenzene 98%
1,3-Dichlorobenzene


Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173 °C
Melting Pt: –25 °C
Density: 1.288
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000573
CAS Number: 541-73-1
EINECS: 208-792-1
UN: 3082
ADR: "9",III
Merck Index: 13,03080
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty