Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid dihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethanedioic acid dihydrate

A13866-0B A13866-0I A13866-0B A13866-30 A13866-0I A13866-30
AAAA13866-0BEA 100.99 USD InStock
AAAA13866-0B CAAAAA13866-0I CAAAAA13866-30 AAAA13866-0I CAAAAA13866-0B AAAA13866-30
Oxalic acid dihydrate 98%
Oxalic acid dihydrate


Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 101…105 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty