Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Sulfolene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-sulfoleneButadiene sulphoneButadiene sulfone3-sulpholene

A13887-0B A13887-22 A13887-0I A13887-0B A13887-0I A13887-36 A13887-22 A13887-36
AAAA13887-0IEA 388.99 USD InStock
CAAAAA13887-0B CAAAAA13887-36 AAAA13887-0I CAAAAA13887-0I AAAA13887-36 AAAA13887-0B CAAAAA13887-22 AAAA13887-22
3-Sulfolene 98%
3-Sulpholene


Formula: C₄H₆O₂S
MW: 118.16 g/mol
Melting Pt: 63…67 °C
Density: 1.314
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005481
CAS Number: 77-79-2
EINECS: 201-059-7
UN: 3335
ADR: "9",
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty