Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Pyridone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Hydroxypyridine 3-Pyridinol β-Pyridone

A13910-18 A13910-0B A13910-22 A13910-36 A13910-22 A13910-36 A13910-18 A13910-0B
AAAA13910-22EA 55.19 USD InStock
CAAAA13910-18 AAAA13910-22 AAA13910-0B CAAAAA13910-22 AAA13910-18 AAAA13910-36 CAAAA13910-0B CAAAAA13910-36
3-Pyridone 98%
3-Pyridone


Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 180 °C/22 mm
Melting Pt: 125…128 °C
Density: 1.2 g/cm³ (–70 °C)
Flash Pt: 173 °C(343 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty