Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hydroxy-L-tryptophan hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Hydroxyl-L-tryptophanL-5-HydroxytryptophanOxitriptan

A13954-03 A13954-03 A13954-01 A13954-01
AAAA13954-01EA 18.48 USD InStock
CAAAAA13954-01 CAAAAA13954-03 AAAA13954-01 AAAA13954-03
5-Hydroxy-L-tryptophan hydrate 98%
5-Hydroxy-L-tryptophan
For routes to 2-bromo-5-hydroxytryptophan derivatives, see: Synthesis, 28 (1996).


Formula: C₁₁H₁₂N₂O₃·xH₂O
MW: 220.23 g/mol (anhy)
Melting Pt: 258 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064341
CAS Number: 4350-09-8
EINECS: 224-411-1
UN: 2811
ADR: "6.1",III
Merck Index: 12,04895
Order Now
Specification Test Results
Optical rotation -33° (c=1 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty