Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) nitrate hemipentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cupric Nitrate HydrateCupric nitrate hemipentahydrateCopper dinitrate hemipentahydrateCupric nitrate 2.5-hydrateCupric nitrate hemi(pentahydrate)

A14004-0I A14004-30 A14004-0B A14004-0B A14004-0I A14004-30
AAAA14004-0IEA 513.4 USD InStock
AAAA14004-0I CAAAAA14004-30 CAAAAA14004-0I CAAAAA14004-0B AAAA14004-0B AAAA14004-30
Copper(II) nitrate hemipentahydrate 98%
Copper (II) nitrate hemipentahydrate


Formula: Cu(NO₃)₂·2.5H₂O
MW: 232.59 g/mol
Boiling Pt: 170 °C (dec.)
Melting Pt: ca 115 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: "5.1",III
Merck Index: 13,02671
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty