Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Diaminobenzoic acid 98% may contain up to 3% moisture

Supplier: Thermo Scientific Chemicals
Warning

A14075-18 A14075-22 A14075-22 A14075-18 A14075-36 A14075-36
AAA14075-18EA 33.69 USD InStock
AAA14075-18 CAAAAA14075-22 AAAA14075-22 CAAAAA14075-36 AAAA14075-36 CAAAA14075-18
3,5-Diaminobenzoic acid 98% may contain up to 3% moisture
3,5-Diaminobenzoic acid


Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 546 °C (1013 hPa)
Melting Pt: ca 235 °C (dec.)
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 210 °C(410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007807
CAS Number: 535-87-5
EINECS: 208-621-0
Merck Index: 13,02997
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty