Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Decanone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Decan-2-one

A14145-06 A14145-14 A14145-22 A14145-06 A14145-14 A14145-22
AAA14145-14EA 87.19 USD InStock
CAAAAA14145-06 AAA14145-14 AAAA14145-06 AAA14145-22 CAAAA14145-14 CAAAA14145-22
2-Decanone 97%
2-Decanone


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 210…212 °C
Melting Pt: 3 °C
Density: 0.824
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009571
CAS Number: 693-54-9
EINECS: 211-752-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty