Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sebacic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: Decanedioic acid1,8-Octanedicarboxylic acid1,10-Decanedioic acid

A14158-30 A14158-0B A14158-0I A14158-0B A14158-22 A14158-0I A14158-30 A14158-22
AAAA14158-0IEA 373.99 USD InStock
CAAAAA14158-0I CAAAAA14158-30 AAAA14158-0I CAAAAA14158-22 AAAA14158-30 AAAA14158-0B CAAAAA14158-0B AAAA14158-22
Sebacic acid 98+%
Sebacic acid


Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 295 °C/100 mm
Melting Pt: 131…134 °C
Density: 1.271
Flash Pt: 220 °C(428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty