Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentanone 99%

Supplier: Thermo Scientific Chemicals
Warning

A14222-AP A14222-0F A14222-AK A14222-AP A14222-AK A14222-0F
AAAA14222-AKEA 57.39 USD InStock
CAAAAA14222-AP AAAA14222-AK AAAA14222-AP CAAAAA14222-0F CAAAAA14222-AK AAAA14222-0F
Cyclopentanone 99%
Cyclopentanone


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 130…131 °C
Melting Pt: –51 °C
Density: 0.949
Flash Pt: 30 °C(86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001409
CAS Number: 120-92-3
EINECS: 204-435-9
UN: 2245
ADR: "3",III
Merck Index: 13,02771
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty