Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethoxyacetylene 50% (w/w) in hexane

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: ethoxy-ethyneethynoxyethaneEthyl ethynyl ether

A14245-14 A14245-06 A14245-03 A14245-14 A14245-03 A14245-06
AAAA14245-06EA 107.99 USD InStock
AAAA14245-06 CAAAAA14245-03 CAAAAA14245-14 AAAA14245-14 AAAA14245-03 CAAAAA14245-06
Ethoxyacetylene 50% (w/w) in hexane
Ethoxyacetylene


Formula: C₄H₆O
MW: 70.091 g/mol
Boiling Pt: 55…56 °C
Density: 0.718
Flash Pt: -28 °C(-18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009247
CAS Number: 927-80-0
EINECS: 213-164-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty