Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Xylidine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,6-Dimethylaniline 2-Amino-m-xylene

A14512-22 A14512-36 A14512-0B A14512-0B A14512-22 A14512-36
AAAA14512-22EA 46.81 USD InStock
AAAA14512-22 CAAAAA14512-36 AAA14512-0B CAAAAA14512-22 AAAA14512-36 CAAAA14512-0B
2,6-Xylidine 99%
2,6-Xylidine


Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 216 °C
Melting Pt: 11 °C
Density: 0.981
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007747
CAS Number: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: "6.1",II
Merck Index: 14,10084
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty