Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maleic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14596-0I A14596-30 A14596-0I A14596-0B A14596-30 A14596-0B
AAAA14596-30EA 40.53 USD InStock
CAAAAA14596-0B CAAAAA14596-30 AAAA14596-30 CAAAAA14596-0I AAAA14596-0I AAAA14596-0B
Maleic acid 98+%
Maleic acid


Formula: C₄H₄O₄
MW: 116.07 g/mol
Melting Pt: 132…136 °C
Density: 1.59
Flash Pt: 127 °C(260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: "8",III
Merck Index: 13,05726
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/21/2297221.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty