Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethyl)phenylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Trifluoromethyl)benzyl cyanide

A14663-14 A14663-18 A14663-14 A14663-18
AAAA14663-18EA 251.99 USD InStock
CAAAA14663-14 AAAA14663-18 CAAAAA14663-18 AAA14663-14
3-(Trifluoromethyl)phenylacetonitrile 97%
3-(Trifluoromethyl)phenylacetonitrile


Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 92…93 °C/4 mm
Density: 1.187
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001913
CAS Number: 2338-76-3
EINECS: 219-053-8
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty